(R)-2-(Boc-aminomethyl)morpholine
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl N-[(2R)-morpholin-2-ylmethyl]carbamate
OR451057-500MG
APOSOR451057-500MGEA
960
EUR
APOSOR451057-500MG
(R)-2-(Boc-aminomethyl)morpholine
(R)-2-(Boc-aminomethyl)morpholine
Formel:
C₁₀H₂₀N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12545850 CAS-Nummer: 186202-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...