(R)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride

Lieferant: Apollo Scientific

OR451058-1G
APOSOR451058-1GEA 873 EUR
APOSOR451058-1G
(R)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride
(R)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride
Formel: C₉H₁₈N₂O·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28975215
CAS-Nummer: 511295-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR