(3S)-3-Phenyl-morpholine hydrochloride

Lieferant: Apollo Scientific

OR451059-1G
APOSOR451059-1GEA 1160 EUR
APOSOR451059-1G
(3S)-3-Phenyl-morpholine hydrochloride
(3S)-3-Phenyl-morpholine hydrochloride
Formel: C₁₀H₁₃NO·ClH
Molecular Weight: 199,677 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12910598
CAS-Nummer: 1432230-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR