(R)-3-Phenylmorpholine hydrochloride

Lieferant: Apollo Scientific

OR451060-1G
APOSOR451060-1GEA 165 EUR
APOSOR451060-1G
(R)-3-Phenylmorpholine hydrochloride
(R)-3-Phenylmorpholine hydrochloride
Formel: C₁₀H₁₃NO·ClH
Molecular Weight: 199,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12910599
CAS-Nummer: 1624261-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR