3-(Methoxymethyl)-morpholine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-(Methoxymethyl)morpholine hydrochloride

OR451061-1G
APOSOR451061-1GEA 549 EUR
APOSOR451061-1G
3-(Methoxymethyl)-morpholine hydrochloride
3-(Methoxymethyl)-morpholine hydrochloride
Formel: C₆H₁₃NO₂·ClH
Molecular Weight: 167,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18425620
CAS-Nummer: 955400-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR