4-Boc-3-(aminomethyl)-morpholine

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-(aminomethyl)morpholine-4-carboxylate

OR451062-1G
APOSOR451062-1GEA 1100 EUR
APOSOR451062-1G
4-Boc-3-(aminomethyl)-morpholine
4-Boc-3-(aminomethyl)-morpholine
Formel: C₁₀H₂₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06738665
CAS-Nummer: 475106-18-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR