(R)-4-Boc-3-(aminomethyl)-morpholine

Lieferant: Apollo Scientific

Synonyme: (R)-tert-Butyl 3-(aminomethyl)morpholine-4-carboxylate

OR451064-1G
APOSOR451064-1GEA 505 EUR
APOSOR451064-1G
(R)-4-Boc-3-(aminomethyl)-morpholine
(R)-4-Boc-3-(aminomethyl)-morpholine
Formel: C₁₀H₂₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06739119
CAS-Nummer: 1187929-33-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR