(S)-N,N-Dimethyl-2-morpholinemethanamine dihydrochloride

Lieferant: Apollo Scientific

OR451065-1G
APOSOR451065-1GEA 1250 EUR
APOSOR451065-1G
(S)-N,N-Dimethyl-2-morpholinemethanamine dihydrochloride
(S)-N,N-Dimethyl-2-morpholinemethanamine dihydrochloride
Formel: C₇H₁₆N₂O·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28016670
CAS-Nummer: 214273-18-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR