(S)-2-(Methoxymethyl)-morpholine hydrochloride

Lieferant: Apollo Scientific

Synonyme: (S)-2-(Methoxymethyl)morpholine hydrochloride

OR451067-1G
APOSOR451067-1GEA 220 EUR
APOSOR451067-1G
(S)-2-(Methoxymethyl)-morpholine hydrochloride
(S)-2-(Methoxymethyl)-morpholine hydrochloride
Formel: C₆H₁₃NO₂·ClH
Molecular Weight: 167.634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16294688
CAS-Nummer: 141196-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR