(R)-2-(Methoxymethyl)-morpholine hydrochloride

Lieferant: Apollo Scientific

Synonyme: (R)-2-(Methoxymethyl)morpholine hydrochloride

OR451068-1G
APOSOR451068-1GEA 136 EUR
APOSOR451068-1G
(R)-2-(Methoxymethyl)-morpholine hydrochloride
(R)-2-(Methoxymethyl)-morpholine hydrochloride
Formel: C₆H₁₃NO₂·ClH
Molecular Weight: 167,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16294689
CAS-Nummer: 141196-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR