Ethyl 3-cyclobutyl-3-oxopropanoate

Lieferant: Apollo Scientific

OR451071-25G
APOSOR451071-25GEA 2700 EUR
APOSOR451071-25G
Ethyl 3-cyclobutyl-3-oxopropanoate
Ethyl 3-cyclobutyl-3-oxopropanoate
Formel: C₉H₁₄O₃
Molecular Weight: 170,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410651
CAS-Nummer: 24922-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR