1-(4-Bromophenyl)-cyclobutanemethanamine

Lieferant: Apollo Scientific

Synonyme: (1-(4-Bromophenyl)cyclobutyl)methanamine

OR451072-1G
APOSOR451072-1GEA 873 EUR
APOSOR451072-1G
1-(4-Bromophenyl)-cyclobutanemethanamine
1-(4-Bromophenyl)-cyclobutanemethanamine
Formel: C₁₁H₁₄BrN
Molecular Weight: 240,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11188929
CAS-Nummer: 1039932-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR