Spiro[cyclobutane-1,3'-indoline] hydrochloride

Lieferant: Apollo Scientific

OR451074-500MG
APOSOR451074-500MGEA 1090 EUR
APOSOR451074-500MG
Spiro[cyclobutane-1,3'-indoline] hydrochloride
Spiro[cyclobutane-1,3'-indoline] hydrochloride
Formel: C₁₁H₁₃N·ClH
Molecular Weight: 195,689 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28023681
CAS-Nummer: 32670-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR