β-Oxo-cyclobutanepropanoic acid 1,1-dimethylethyl ester

Lieferant: Apollo Scientific

OR451076-1G
APOSOR451076-1GEA 960 EUR
APOSOR451076-1G
β-Oxo-cyclobutanepropanoic acid 1,1-dimethylethyl ester
β-Oxo-cyclobutanepropanoic acid 1,1-dimethylethyl ester
Formel: C₁₁H₁₈O₃
Molecular Weight: 198,259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11974506
CAS-Nummer: 134302-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR