(2R)-2-Amino-2-cyclobutylethan-1-ol hydrochloride

Lieferant: Apollo Scientific

OR451077-500MG
APOSOR451077-500MGEA 1320 EUR
APOSOR451077-500MG
(2R)-2-Amino-2-cyclobutylethan-1-ol hydrochloride
(2R)-2-Amino-2-cyclobutylethan-1-ol hydrochloride
Formel: C₆H₁₃NO·ClH
Molecular Weight: 151,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975201
CAS-Nummer: 1259608-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR