1-(Cyclopropylcarbonyl)piperazine

Lieferant: Apollo Scientific

OR451081-25G
APOSOR451081-25GEA 150 EUR
APOSOR451081-25G
1-(Cyclopropylcarbonyl)piperazine
1-(Cyclopropylcarbonyl)piperazine
Formel: C₈H₁₄N₂O
Molecular Weight: 154,213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06369645
CAS-Nummer: 59878-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR