D-Cyclopropylalinine

Lieferant: Apollo Scientific

Synonyme: β-Cyclopropyl-D-Ala-OH, H-β-Cyclopropyl-D-Ala-OH

OR451082-5G
APOSOR451082-5GEA 242 EUR
APOSOR451082-5G
D-Cyclopropylalinine
D-Cyclopropylalinine
Formel: C₆H₁₁NO₂
Molecular Weight: 129,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02259520
CAS-Nummer: 121786-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR