1-(2,4-Dichloro-phenyl)-cyclopropylamine

Lieferant: Apollo Scientific

OR451085-1G
APOSOR451085-1GEA 771 EUR
APOSOR451085-1G
1-(2,4-Dichloro-phenyl)-cyclopropylamine
1-(2,4-Dichloro-phenyl)-cyclopropylamine
Formel: C₉H₉Cl₂N
Molecular Weight: 202,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371522
CAS-Nummer: 864263-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR