N-(Cyclopropylmethyl)-benzylamine hydrochloride

Lieferant: Apollo Scientific

OR451086-5G
APOSOR451086-5GEA 806 EUR
APOSOR451086-5G
N-(Cyclopropylmethyl)-benzylamine hydrochloride
N-(Cyclopropylmethyl)-benzylamine hydrochloride
Formel: C₁₁H₁₅N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD17214588
CAS-Nummer: 908843-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR