N-(Cyclopropylmethyl)-4-methoxy-benzylamine
Lieferant: Apollo Scientific
OR451087-1G
APOSOR451087-1GEA
1160
EUR
APOSOR451087-1G
N-(Cyclopropylmethyl)-4-methoxy-benzylamine
N-(Cyclopropylmethyl)-4-methoxy-benzylamine
Formel:
C₁₂H₁₇NO Molecular Weight: 191,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11140261 CAS-Nummer: 1019611-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...