N-(Cyclopropylmethyl)-4-chloro-benzylamine

Lieferant: Apollo Scientific

OR451088-1G
APOSOR451088-1GEA 1160 EUR
APOSOR451088-1G
N-(Cyclopropylmethyl)-4-chloro-benzylamine
N-(Cyclopropylmethyl)-4-chloro-benzylamine
Formel: C₁₁H₁₄ClN
Molecular Weight: 195,689 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11140255
CAS-Nummer: 1019506-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR