N-(Cyclopropylmethyl)-3-bromo-benzylamine

Lieferant: Apollo Scientific

OR451089-1G
APOSOR451089-1GEA 510 EUR
APOSOR451089-1G
N-(Cyclopropylmethyl)-3-bromo-benzylamine
N-(Cyclopropylmethyl)-3-bromo-benzylamine
Formel: C₁₁H₁₄BrN
Molecular Weight: 240,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11140259
CAS-Nummer: 1019506-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR