N-(Cyclopropylmethyl)-2-chloro-benzylamine
Lieferant: Apollo Scientific
OR451090-1G
APOSOR451090-1GEA
1160
EUR
APOSOR451090-1G
N-(Cyclopropylmethyl)-2-chloro-benzylamine
N-(Cyclopropylmethyl)-2-chloro-benzylamine
Formel:
C₁₁H₁₄ClN Molecular Weight: 195,689 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11140251 CAS-Nummer: 1019506-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...