N-(Cyclopropylmethyl)-3-methoxy-benzylamine

Lieferant: Apollo Scientific

OR451091-1G
APOSOR451091-1GEA 1160 EUR
APOSOR451091-1G
N-(Cyclopropylmethyl)-3-methoxy-benzylamine
N-(Cyclopropylmethyl)-3-methoxy-benzylamine
Formel: C₁₂H₁₇NO
Molecular Weight: 191,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11140267
CAS-Nummer: 1019506-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR