N-Cyclopropyl-benzylamine

Lieferant: Apollo Scientific

OR451092-5G
APOSOR451092-5GEA 0 EUR
APOSOR451092-5G
N-Cyclopropyl-benzylamine
N-Cyclopropyl-benzylamine
Formel: C₁₀H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00089909
CAS-Nummer: 13324-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR