2-(4-Methylpiperazin-1-ylmethyl)benzoic acid
Lieferant: Apollo Scientific
OR451094-5G
APOSOR451094-5GEA
1540
EUR
APOSOR451094-5G
2-(4-Methylpiperazin-1-ylmethyl)benzoic acid
2-(4-Methylpiperazin-1-ylmethyl)benzoic acid
Formel:
C₁₃H₁₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797777 CAS-Nummer: 514209-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...