1-Boc-2-(1-methylethyl)-piperazine

Lieferant: Apollo Scientific

Synonyme: 1-Boc-2-Isopropylpiperazine

OR451096-1G
APOSOR451096-1GEA 243 EUR
APOSOR451096-1G
1-Boc-2-(1-methylethyl)-piperazine
1-Boc-2-(1-methylethyl)-piperazine
Formel: C₁₂H₂₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08273957
CAS-Nummer: 886766-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR