1-Boc-4-[(2R)-2-aminopropyl]-piperazine dihydrochloride

Lieferant: Apollo Scientific

OR451097-1G
APOSOR451097-1GEA 771 EUR
APOSOR451097-1G
1-Boc-4-[(2R)-2-aminopropyl]-piperazine dihydrochloride
1-Boc-4-[(2R)-2-aminopropyl]-piperazine dihydrochloride
Formel: C₁₂H₂₅N₃O₂·2ClH
Molecular Weight: 316,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975202
CAS-Nummer: 1303975-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR