1-Boc-4-[(2S)-2-aminopropyl]-piperazine dihydrochloride

Lieferant: Apollo Scientific

OR451098-1G
APOSOR451098-1GEA 771 EUR
APOSOR451098-1G
1-Boc-4-[(2S)-2-aminopropyl]-piperazine dihydrochloride
1-Boc-4-[(2S)-2-aminopropyl]-piperazine dihydrochloride
Formel: C₁₂H₂₅N₃O₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28975218
CAS-Nummer: 1017606-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR