(R)-1-Boc-3-isopropylpiperazin

Lieferant: Apollo Scientific

OR451099-5G
APOSOR451099-5GEA 717 EUR
APOSOR451099-5G
(R)-1-Boc-3-isopropylpiperazin
(R)-1-Boc-3-isopropylpiperazin
Formel: C₁₂H₂₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07772113
CAS-Nummer: 928025-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR