(R)-2-Benzylpiperazine dihydrochloride

Lieferant: Apollo Scientific

OR451100-1G
APOSOR451100-1GEA 261 EUR
APOSOR451100-1G
(R)-2-Benzylpiperazine dihydrochloride
(R)-2-Benzylpiperazine dihydrochloride
Formel: C₁₁H₁₆N₂·2ClH
Molecular Weight: 249,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954327
CAS-Nummer: 131288-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR