(S)-4-Benzyl-2-hydroxymethylpiperazine
Lieferant: Apollo Scientific
OR451102-1G
APOSOR451102-1GEA
1540
EUR
APOSOR451102-1G
(S)-4-Benzyl-2-hydroxymethylpiperazine
(S)-4-Benzyl-2-hydroxymethylpiperazine
Formel:
C₁₂H₁₈N₂O Molecular Weight: 206,284 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11113047 CAS-Nummer: 149715-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...