(S)-1-Boc-2-phenylpiperazine

Lieferant: Apollo Scientific

Synonyme: (S)-tert-Butyl 2-phenylpiperazine-1-carboxylate

OR451104-1G
APOSOR451104-1GEA 296 EUR
APOSOR451104-1G
(S)-1-Boc-2-phenylpiperazine
(S)-1-Boc-2-phenylpiperazine
Formel: C₁₅H₂₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08685943
CAS-Nummer: 1240583-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR