(R)-tert-Butyl 2-phenylpiperazine-1-carboxylate

Lieferant: Apollo Scientific
Achtung

OR451105-1G
APOSOR451105-1GEA 264 EUR
APOSOR451105-1G
(R)-tert-Butyl 2-phenylpiperazine-1-carboxylate
(R)-tert-Butyl 2-phenylpiperazine-1-carboxylate
Formel: C₁₅H₂₂N₂O₂
Molecular Weight: 262,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972241
CAS-Nummer: 859518-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR