(S)-1-Boc-3-Phenylpiperazine

Lieferant: Apollo Scientific

Synonyme: (S)-tert-Butyl 3-phenylpiperazine-1-carboxylate

OR451106-1G
APOSOR451106-1GEA 960 EUR
APOSOR451106-1G
(S)-1-Boc-3-Phenylpiperazine
(S)-1-Boc-3-Phenylpiperazine
Formel: C₁₅H₂₂N₂O₂
Molecular Weight: 262.347 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08685941
CAS-Nummer: 1221274-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR