a-Cyclopropyl-4-bromo-benzylamine

Lieferant: Apollo Scientific

OR451108-1G
APOSOR451108-1GEA 1160 EUR
APOSOR451108-1G
a-Cyclopropyl-4-bromo-benzylamine
a-Cyclopropyl-4-bromo-benzylamine
Formel: C₁₀H₁₂BrN
Lagertemperatur: Raumtemperatur
MDL: MFCD05192037
CAS-Nummer: 90868-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR