2-Amino-2-cyclopropylethan-1-ol hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-2-cyclopropylethanol hydrochloride

OR451109-1G
APOSOR451109-1GEA 678 EUR
APOSOR451109-1G
2-Amino-2-cyclopropylethan-1-ol hydrochloride
2-Amino-2-cyclopropylethan-1-ol hydrochloride
Formel: C₅H₁₁NO·ClH
Molecular Weight: 137,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18839092
CAS-Nummer: 1306603-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR