4-Cyclopropylphenylacetic acid

Lieferant: Apollo Scientific
Achtung

OR451113-1G
APOSOR451113-1GEA 654 EUR
APOSOR451113-1G
4-Cyclopropylphenylacetic acid
4-Cyclopropylphenylacetic acid
Formel: C₁₁H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18452123
CAS-Nummer: 40641-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR