4-(Cyclopropylmethoxy)-benzoic acid

Lieferant: Apollo Scientific

Synonyme: 4-(Cyclopropylmethoxy)benzoic acid

OR451114-5G
APOSOR451114-5GEA 1650 EUR
APOSOR451114-5G
4-(Cyclopropylmethoxy)-benzoic acid
4-(Cyclopropylmethoxy)-benzoic acid
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11156580
CAS-Nummer: 355391-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR