Fmoc-L-cyclopropylalanine
Lieferant: Apollo Scientific
Synonyme:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopropylpropanoic acid, Fmoc-β-cyclopropyl-L-Alanine, Fmoc-β-cyclopropyl-Ala-OH
OR451117-1G
APOSOR451117-1GEA
77
EUR
APOSOR451117-1G
Fmoc-L-cyclopropylalanine
Fmoc-L-cyclopropylalanine
Formel:
C₂₁H₂₁NO₄ Molecular Weight: 351.402 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798640 CAS-Nummer: 214750-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...