N-Boc-(2-Cyclopropyl-2-hydroxyethyl)amine

Lieferant: Apollo Scientific

OR451120-1G
APOSOR451120-1GEA 1320 EUR
APOSOR451120-1G
N-Boc-(2-Cyclopropyl-2-hydroxyethyl)amine
N-Boc-(2-Cyclopropyl-2-hydroxyethyl)amine
Formel: C₁₀H₁₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11977306
CAS-Nummer: 121102-91-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR