1-Boc-4-cyano-4-(cyclopropylmethyl)-piperidine

Lieferant: Apollo Scientific

OR451121-1G
APOSOR451121-1GEA 1320 EUR
APOSOR451121-1G
1-Boc-4-cyano-4-(cyclopropylmethyl)-piperidine
1-Boc-4-cyano-4-(cyclopropylmethyl)-piperidine
Formel: C₁₅H₂₄N₂O₂
Molecular Weight: 264,363 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11849800
CAS-Nummer: 895132-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR