(3,3-Dimethoxycyclobutyl)methanol

Lieferant: Apollo Scientific

OR451123-1G OR451123-10MG OR451123-250MG OR451123-500MG OR451123-2.5G
APOSOR451123-1GEA 171 EUR
APOSOR451123-1G APOSOR451123-10MG APOSOR451123-250MG APOSOR451123-500MG APOSOR451123-2.5G
(3,3-Dimethoxycyclobutyl)methanol
(3,3-Dimethoxycyclobutyl)methanol
Formel: C₇H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11110681
CAS-Nummer: 175021-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR