1-(4-Methylphenyl)cyclobutanecarboxylic acid

Lieferant: Apollo Scientific

OR451124-1G
APOSOR451124-1GEA 873 EUR
APOSOR451124-1G
1-(4-Methylphenyl)cyclobutanecarboxylic acid
1-(4-Methylphenyl)cyclobutanecarboxylic acid
Formel: C₁₂H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11037068
CAS-Nummer: 50921-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR