1-(Aminomethyl)-cyclobutanecarboxylic acid hydrochloride

Lieferant: Apollo Scientific

OR451126-1G
APOSOR451126-1GEA 241 EUR
APOSOR451126-1G
1-(Aminomethyl)-cyclobutanecarboxylic acid hydrochloride
1-(Aminomethyl)-cyclobutanecarboxylic acid hydrochloride
Formel: C₆H₁₁NO₂·ClH
Molecular Weight: 165,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21602353
CAS-Nummer: 1360547-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR