1-(3-Chlorophenyl)-cyclobutanemethanamine
Lieferant: Apollo Scientific
Synonyme:
(1-(3-Chlorophenyl)cyclobutyl)methanamine
OR451128-1G
APOSOR451128-1GEA
960
EUR
APOSOR451128-1G
1-(3-Chlorophenyl)-cyclobutanemethanamine
1-(3-Chlorophenyl)-cyclobutanemethanamine
Formel:
C₁₁H₁₄ClN Molecular Weight: 195,689 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09904043 CAS-Nummer: 115816-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...