(R)-2-Cyclobutylglycine

Lieferant: Apollo Scientific

OR451130-1G
APOSOR451130-1GEA 951 EUR
APOSOR451130-1G
(R)-2-Cyclobutylglycine
(R)-2-Cyclobutylglycine
Formel: C₆H₁₁NO₂
Molecular Weight: 129,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436201
CAS-Nummer: 49607-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR