3-Cyclobutyl-3-oxo-propionitrile
Lieferant: Apollo Scientific
Synonyme:
3-Cyclobutyl-3-oxopropanenitrile, b-Oxo-cyclobutanepropanenitrile
OR451133-1G
APOSOR451133-1GEA
579
EUR
APOSOR451133-1G
3-Cyclobutyl-3-oxo-propionitrile
3-Cyclobutyl-3-oxo-propionitrile
Formel:
C₇H₉NO Molecular Weight: 123,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787182 CAS-Nummer: 118431-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...