1-Pyrrolidin-3-yl-1H-pyrazole dihydrochloride

Lieferant: Apollo Scientific

OR451136-1G
APOSOR451136-1GEA 688 EUR
APOSOR451136-1G
1-Pyrrolidin-3-yl-1H-pyrazole dihydrochloride
1-Pyrrolidin-3-yl-1H-pyrazole dihydrochloride
Formel: C₇H₁₃Cl₂N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16039347
CAS-Nummer: 1242339-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR