4-Ethynyl-1H-pyrazole 97%

Lieferant: Apollo Scientific

OR451137-1G
APOSOR451137-1GEA 190 EUR
APOSOR451137-1G
4-Ethynyl-1H-pyrazole 97%
4-Ethynyl-1H-pyrazole
Formel: C₅H₄N₂
Molecular Weight: 92,099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18207413
CAS-Nummer: 57121-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR